The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001005743.1 | NP_001005743.1 | 5 - 6 | 525 | 113 |
NM_001005744.1 | NP_001005744.1 | 5 - 6 | 525 | 113 | |
NM_001320114.1 | NP_001307043.1 | 5 - 6 | 545 | 113 | |
GenBank mRNA | AF015040.1 | - | 5 - 6 | 483 | 113 |
AF171938.1 | - | 5 - 6 | 475 | 113 | |
AF171939.1 | - | 5 - 6 | 475 | 113 | |
AK304193.1 | - | 3 - 4 | 328 | 113 | |
BX248073.1 | - | 4 - 5 | 382 | 113 | |
EU265734.1 | - | 3 - 4 | 244 | 113 | |
EU265736.1 | - | 3 - 4 | 244 | 113 |