The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_052877.4 | NP_443109.2 | 2 - 3 | 175 | 72 |
NM_201542.4 | NP_963836.2 | 2 - 3 | 175 | 72 | |
GenBank mRNA | AF521562.1 | - | 1 - 2 | 127 | 72 |
AV684532.1 | - | 2 - 3 | 212 | 72 | |
BC010543.1 | - | 1 - 2 | 116 | 72 | |
BG620658.1 | - | 2 - 3 | 176 | 72 | |
BI488866.1 | - | 1 - 2 | 131 | 72 |