The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001303429.1 | NP_001290358.1 | 6 - 7 | 1517 | 140 |
GenBank mRNA | AF028785.1 | - | 6 - 7 | 1452 | 140 |