The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001166.4 | NP_001157.1 | 7 - 8 | 3073 | 119 |
NM_001256163.1 | NP_001243092.1 | 7 - 8 | 3350 | 119 | |
NM_001256166.1 | NP_001243095.1 | 7 - 8 | 1670 | 119 | |
GenBank mRNA | AK225856.1 | - | 6 - 7 | 2065 | 119 |
AK303197.1 | - | 7 - 8 | 1638 | 119 | |
AK313446.1 | - | 6 - 7 | 1878 | 119 | |
BC016174.1 | - | 6 - 7 | 1649 | 119 | |
BC028578.1 | - | 7 - 8 | 3351 | 119 | |
BX647978.1 | - | 7 - 8 | 3020 | 119 | |
L49431.1 | - | 6 - 7 | 1815 | 119 | |
U37547.1 | - | 6 - 7 | 2780 | 119 | |
U45879.1 | - | 6 - 7 | 1858 | 119 |