The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001243533.1 | NP_001230462.1 | 1 - 2 | 136 | 82 |
GenBank mRNA | AF411102.1 | - | 1 - 2 | 76 | 82 |
BC028713.1 | - | 1 - 2 | 78 | 82 |
Gene Name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromosome | |
---|---|---|---|---|---|---|
Upstream | tripartite motif containing 63 | TRIM63 | IRF, MURF1, MURF2, RNF28, SMRZ | -44.1kb | Anti-sense | Chr. - 26051304 - 26067634 on Build GRCh38 |
Downstream | family with sequence similarity 110 member D | FAM110D | GRRP1 | 46.4kb | Sense | Chr. - 26159020 - 26162628 on Build GRCh38 |