The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001204519.1 | NP_001191448.1 | 11 - 12 | 1659 | 83 |
NM_001204520.1 | NP_001191449.1 | 10 - 11 | 1590 | 83 | |
NM_002561.3 | NP_002552.2 | 11 - 12 | 1662 | 83 | |
NM_175080.2 | NP_778255.1 | 10 - 11 | 1587 | 83 | |
GenBank mRNA | AF016709.1 | - | 11 - 12 | 1299 | 83 |
AF070573.1 | - | 10 - 11 | 1215 | 83 | |
AK290889.1 | - | 11 - 12 | 1662 | 83 | |
AK293610.1 | - | 9 - 10 | 1262 | 83 | |
AK307959.1 | - | 11 - 12 | 1492 | 83 | |
BC039015.1 | - | 11 - 12 | 1403 | 83 | |
DQ851492.1 | - | 4 - 5 | 493 | 83 | |
U49395.1 | - | 11 - 12 | 1312 | 83 | |
U49396.1 | - | 10 - 11 | 1240 | 83 |