Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GRXCR2
Entrez Gene ID
643226
Gene Name
glutaredoxin and cysteine rich domain containing 2
Gene Aliases
DFNB101
Chromosome Location
Chr.5: 145858538 - 145931673 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001080516.1 NP_001073985.1 1 - 2 338 144
GenBank mRNA DN831963.1 - 1 - 2 276 144

View Full Product Details