Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- EHBP1
- Entrez Gene ID
-
23301
- Gene Name
- EH domain binding protein 1
- Gene Aliases
-
HPC12, NACSIN
- Chromosome Location
-
Chr.2: 62673851 - 63046487 on Build GRCh38
View Full Product Details