The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NR_033519.1 | 1 - 2 | 394 | 102 | |
XM_017006870.1 | XP_016862359.1 | 1 - 2 | 592 | 102 | |
GenBank mRNA | AK304334.1 | - | 1 - 2 | 104 | 102 |
BX647237.1 | - | 1 - 2 | 109 | 102 |