The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002214.2 | NP_002205.1 | 12 - 13 | 2721 | 122 |
XM_011515393.2 | XP_011513695.1 | 13 - 14 | 2524 | 122 | |
XM_011515394.2 | XP_011513696.1 | 13 - 14 | 2122 | 122 | |
XM_011515396.1 | XP_011513698.1 | 13 - 14 | 2793 | 122 | |
XM_017012178.1 | XP_016867667.1 | 13 - 14 | 2418 | 122 | |
XM_017012179.1 | XP_016867668.1 | 13 - 14 | 2877 | 122 | |
XM_017012180.1 | XP_016867669.1 | 13 - 14 | 3170 | 122 | |
XM_017012181.1 | XP_016867670.1 | 14 - 15 | 2989 | 122 | |
XM_017012182.1 | XP_016867671.1 | 13 - 14 | 2104 | 122 | |
XM_017012183.1 | XP_016867672.1 | 13 - 14 | 2061 | 122 | |
GenBank mRNA | AK295044.1 | - | 13 - 14 | 2793 | 122 |
M73780.1 | - | 12 - 13 | 2696 | 122 |