The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_033197.2 | NP_149974.2 | 3 - 4 | 82 | |
GenBank mRNA | AF364078.1 | - | 3 - 4 | 82 | |
AK290243.1 | - | 3 - 4 | 82 | ||
AK310964.1 | - | 3 - 4 | 346 | 82 | |
AK313176.1 | - | 3 - 4 | 82 | ||
AY358595.1 | - | 3 - 4 | 82 | ||
BC008429.1 | - | 3 - 4 | 337 | 82 |