The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_020482.5 | NP_065228.4 | 1 - 2 | 200 | 72 |
GenBank mRNA | AK058175.1 | - | 1 - 2 | 166 | 72 |
AK223539.1 | - | 1 - 2 | 84 | 72 | |
BC029490.1 | - | 1 - 2 | 151 | 72 | |
HY020942.1 | - | 1 - 2 | 201 | 72 |