Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MAG
Entrez Gene ID
4099
Gene Name
myelin associated glycoprotein
Gene Aliases
GMA, S-MAG, SIGLEC-4A, SIGLEC4A, SPG75
Chromosome Location
Chr.19: 35292086 - 35313807 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_002361.3 NP_002352.1 1 - 2 122 87
NM_080600.2 NP_542167.1 1 - 2 122 87
GenBank mRNA AK222649.1 - 1 - 2 47 87
AK223562.1 - 1 - 2 83 87
BC093045.1 - 1 - 2 74 87
BI755065.1 - 1 - 2 72 87
DC331168.1 - 1 - 2 122 87

View Full Product Details