The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001199216.1 | NP_001186145.1 | 6 - 7 | 1156 | 80 |
NM_002361.3 | NP_002352.1 | 6 - 7 | 1169 | 80 | |
NM_080600.2 | NP_542167.1 | 6 - 7 | 1169 | 80 | |
GenBank mRNA | AK094545.1 | - | 7 - 8 | 1015 | 80 |
AK222649.1 | - | 6 - 7 | 1094 | 80 | |
AK223562.1 | - | 6 - 7 | 1130 | 80 | |
AK294644.1 | - | 6 - 7 | 1063 | 80 | |
AK308976.1 | - | 6 - 7 | 1081 | 80 | |
BC053347.1 | - | 6 - 7 | 1061 | 80 | |
BC093045.1 | - | 6 - 7 | 1121 | 80 | |
M29273.1 | - | 6 - 7 | 1091 | 80 |