The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000760.3 | NP_000751.1 | 9 - 10 | 1656 | 56 |
NM_156039.3 | NP_724781.1 | 9 - 10 | 1656 | 56 | |
NM_172313.2 | NP_758519.1 | 9 - 10 | 1656 | 56 | |
XM_005270493.1 | XP_005270550.1 | 9 - 10 | 1656 | 56 | |
XM_011540748.2 | XP_011539050.1 | 8 - 9 | 1181 | 56 | |
XM_011540749.1 | XP_011539051.1 | 9 - 10 | 1656 | 56 | |
XM_011540750.1 | XP_011539052.1 | 4 - 5 | 486 | 56 | |
XM_017000370.1 | XP_016855859.1 | 8 - 9 | 1218 | 56 | |
GenBank mRNA | BC053585.1 | - | 9 - 10 | 1249 | 56 |
BX370109.2 | - | 4 - 5 | 477 | 56 | |
DQ424902.1 | - | 7 - 8 | 1072 | 56 | |
M59818.1 | - | 9 - 10 | 1241 | 56 | |
M59819.1 | - | 9 - 10 | 1241 | 56 | |
M59820.1 | - | 9 - 10 | 1241 | 56 | |
X55720.1 | - | 9 - 10 | 1236 | 56 | |
X55721.1 | - | 9 - 10 | 1237 | 56 |