Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
KLHL12
Entrez Gene ID
59349
Gene Name
kelch like family member 12
Gene Aliases
C3IP1, DKIR
Chromosome Location
Chr.1: 202891096 - 202928644 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001303109.1 NP_001290038.1 1 - 2 185 119
NM_021633.3 NP_067646.1 1 - 2 185 119
XM_017001995.1 XP_016857484.1 1 - 2 154 119
GenBank mRNA AF190900.1 - 1 - 2 109 119
AF306689.1 - 1 - 2 154 119
AK027656.1 - 1 - 2 156 119
AK223072.1 - 1 - 2 121 118
BC003183.1 - 1 - 2 137 119
BC004175.2 - 1 - 2 137 119
DB461868.1 - 1 - 2 187 119

View Full Product Details