The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001303051.1 | NP_001289980.1 | 9 - 10 | 1501 | 89 |
NM_001303109.1 | NP_001290038.1 | 7 - 8 | 1218 | 89 | |
NM_021633.3 | NP_067646.1 | 9 - 10 | 1521 | 89 | |
XM_011509835.1 | XP_011508137.1 | 10 - 11 | 1541 | 89 | |
XM_011509836.1 | XP_011508138.1 | 10 - 11 | 1541 | 89 | |
XM_011509837.1 | XP_011508139.1 | 8 - 9 | 1238 | 89 | |
XM_017001995.1 | XP_016857484.1 | 9 - 10 | 1490 | 89 | |
GenBank mRNA | AF190900.1 | - | 9 - 10 | 1445 | 89 |
AF306687.1 | - | 6 - 7 | 795 | 89 | |
AF306689.1 | - | 7 - 8 | 1187 | 89 | |
AK024412.1 | - | 4 - 5 | 612 | 89 | |
AK027656.1 | - | 9 - 10 | 1492 | 89 | |
AK223072.1 | - | 9 - 10 | 1457 | 89 | |
AK301791.1 | - | 9 - 10 | 1456 | 89 | |
BC003183.1 | - | 9 - 10 | 1473 | 89 | |
BC004175.2 | - | 9 - 10 | 1473 | 89 |