Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- NIPA2
- Entrez Gene ID
-
81614
- Gene Name
- non imprinted in Prader-Willi/Angelman syndrome 2
- Gene Aliases
-
-
- Chromosome Location
-
Chr.15: 22838641 - 22868384 on Build GRCh38
View Full Product Details