Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SLFN13
Entrez Gene ID
146857
Gene Name
schlafen family member 13
Gene Aliases
SLFN10
Chromosome Location
Chr.17: 35435096 - 35448837 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_144682.5 NP_653283.3 3 - 4 1336 75
XM_005257922.2 XP_005257979.1 3 - 4 1317 75
XM_011524383.2 XP_011522685.1 3 - 4 292 75
XM_017024232.1 XP_016879721.1 2 - 3 2858 75
XM_017024233.1 XP_016879722.1 3 - 4 2344 75
GenBank mRNA AK056514.1 - 4 - 5 310 75
AK122750.1 - 4 - 5 665 75
AK126184.1 - 1 - 2 378 75
AK127728.1 - 1 - 2 1389 75
AL832726.1 - 3 - 4 1267 75
BC136622.1 - 3 - 4 1237 75
BC171771.1 - 3 - 4 1237 75
CR749630.1 - 3 - 4 1253 75

View Full Product Details