Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TBC1D5
Entrez Gene ID
9779
Gene Name
TBC1 domain family member 5
Gene Aliases
-
Chromosome Location
Chr.3: 17157162 - 17742739 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001134381.1 NP_001127853.1 15 - 16 1394 93
NM_014744.2 NP_055559.1 14 - 15 2795 93
XM_005265611.1 XP_005265668.1 14 - 15 1365 93
XM_005265612.1 XP_005265669.1 13 - 14 1300 93
XM_005265614.1 XP_005265671.1 14 - 15 1365 93
XM_005265615.1 XP_005265672.1 13 - 14 1295 93
XM_005265616.4 XP_005265673.1 12 - 13 1224 93
XM_006713430.1 XP_006713493.1 14 - 15 1415 93
XM_011534281.1 XP_011532583.1 16 - 17 1676 93
XM_011534283.2 XP_011532585.1 15 - 16 1454 93
XM_011534284.2 XP_011532586.1 15 - 16 1499 93
XM_011534286.1 XP_011532588.1 14 - 15 1431 93
XM_011534287.1 XP_011532589.1 14 - 15 1413 93
XM_017007552.1 XP_016863041.1 17 - 18 1777 93
XM_017007553.1 XP_016863042.1 17 - 18 1807 93
XM_017007554.1 XP_016863043.1 22 - 23 2420 93
XM_017007555.1 XP_016863044.1 16 - 17 1659 93
XM_017007556.1 XP_016863045.1 15 - 16 1639 93
XM_017007557.1 XP_016863046.1 13 - 14 1259 93
XM_017007558.1 XP_016863047.1 13 - 14 1289 93
XM_017007559.1 XP_016863048.1 15 - 16 1639 93
XM_017007560.1 XP_016863049.1 15 - 16 1469 93
XM_017007561.1 XP_016863050.1 13 - 14 1300 93
XM_017007562.1 XP_016863051.1 14 - 15 1413 93
XM_017007563.1 XP_016863052.1 12 - 13 1219 93
XM_017007564.1 XP_016863053.1 13 - 14 1295 93
XM_017007565.1 XP_016863054.1 14 - 15 1593 93
XM_017007566.1 XP_016863055.1 14 - 15 1382 93
XM_017007567.1 XP_016863056.1 12 - 13 1223 93
XM_017007568.1 XP_016863057.1 13 - 14 1259 93
GenBank mRNA AB449886.1 - 12 - 13 1130 93
AK309374.1 - 10 - 11 1055 93
AK310539.1 - 15 - 16 1394 93
BC013145.1 - 13 - 14 1307 93
D86965.1 - 14 - 15 2794 93

View Full Product Details