Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TBC1D5
Entrez Gene ID
9779
Gene Name
TBC1 domain family member 5
Gene Aliases
-
Chromosome Location
Chr.3: 17157162 - 17742739 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001134381.1 NP_001127853.1 21 - 22 2180 101
NM_014744.2 NP_055559.1 19 - 20 3515 101
XM_005265611.1 XP_005265668.1 20 - 21 2151 101
XM_005265612.1 XP_005265669.1 19 - 20 2086 101
XM_005265614.1 XP_005265671.1 19 - 20 2085 101
XM_005265615.1 XP_005265672.1 19 - 20 2081 101
XM_005265616.4 XP_005265673.1 18 - 19 2010 101
XM_006713430.1 XP_006713493.1 20 - 21 2201 101
XM_011534281.1 XP_011532583.1 22 - 23 2462 101
XM_011534283.2 XP_011532585.1 21 - 22 2240 101
XM_011534284.2 XP_011532586.1 21 - 22 2285 101
XM_011534286.1 XP_011532588.1 20 - 21 2217 101
XM_011534287.1 XP_011532589.1 20 - 21 2199 101
XM_017007552.1 XP_016863041.1 23 - 24 2563 101
XM_017007553.1 XP_016863042.1 23 - 24 2593 101
XM_017007554.1 XP_016863043.1 28 - 29 3206 101
XM_017007555.1 XP_016863044.1 22 - 23 2445 101
XM_017007556.1 XP_016863045.1 21 - 22 2425 101
XM_017007557.1 XP_016863046.1 18 - 19 1979 101
XM_017007558.1 XP_016863047.1 18 - 19 2009 101
XM_017007559.1 XP_016863048.1 20 - 21 2359 101
XM_017007560.1 XP_016863049.1 20 - 21 2189 101
XM_017007561.1 XP_016863050.1 18 - 19 2020 101
XM_017007562.1 XP_016863051.1 20 - 21 2199 101
XM_017007563.1 XP_016863052.1 17 - 18 1939 101
XM_017007564.1 XP_016863053.1 18 - 19 2015 101
XM_017007565.1 XP_016863054.1 19 - 20 2313 101
XM_017007566.1 XP_016863055.1 19 - 20 2102 101
XM_017007567.1 XP_016863056.1 17 - 18 1943 101
GenBank mRNA AB449886.1 - 17 - 18 1850 101
AK097990.1 - 6 - 7 959 101
AK310539.1 - 21 - 22 2180 101
BC013145.1 - 18 - 19 2027 101
D86965.1 - 19 - 20 3514 101

View Full Product Details