Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TBC1D5
Entrez Gene ID
9779
Gene Name
TBC1 domain family member 5
Gene Aliases
-
Chromosome Location
Chr.3: 17157162 - 17742739 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001134381.1 NP_001127853.1 6 - 7 537 151
NM_014744.2 NP_055559.1 5 - 6 1938 151
XM_005265611.1 XP_005265668.1 5 - 6 508 151
XM_005265612.1 XP_005265669.1 4 - 5 443 151
XM_005265614.1 XP_005265671.1 5 - 6 508 151
XM_005265615.1 XP_005265672.1 4 - 5 438 151
XM_005265616.4 XP_005265673.1 3 - 4 367 151
XM_006713430.1 XP_006713493.1 5 - 6 558 151
XM_011534281.1 XP_011532583.1 7 - 8 819 151
XM_011534283.2 XP_011532585.1 6 - 7 597 151
XM_011534284.2 XP_011532586.1 6 - 7 642 151
XM_011534286.1 XP_011532588.1 5 - 6 574 151
XM_011534287.1 XP_011532589.1 5 - 6 556 151
XM_017007552.1 XP_016863041.1 8 - 9 920 151
XM_017007553.1 XP_016863042.1 8 - 9 950 151
XM_017007554.1 XP_016863043.1 13 - 14 1563 151
XM_017007555.1 XP_016863044.1 7 - 8 802 151
XM_017007556.1 XP_016863045.1 6 - 7 782 151
XM_017007557.1 XP_016863046.1 4 - 5 402 151
XM_017007558.1 XP_016863047.1 4 - 5 432 151
XM_017007559.1 XP_016863048.1 6 - 7 782 151
XM_017007560.1 XP_016863049.1 6 - 7 612 151
XM_017007561.1 XP_016863050.1 4 - 5 443 151
XM_017007562.1 XP_016863051.1 5 - 6 556 151
XM_017007563.1 XP_016863052.1 3 - 4 362 151
XM_017007564.1 XP_016863053.1 4 - 5 438 151
XM_017007565.1 XP_016863054.1 5 - 6 736 151
XM_017007566.1 XP_016863055.1 5 - 6 525 151
XM_017007567.1 XP_016863056.1 3 - 4 366 151
XM_017007568.1 XP_016863057.1 4 - 5 402 151
GenBank mRNA AB449886.1 - 3 - 4 273 151
AK309374.1 - 1 - 2 198 151
AK310539.1 - 6 - 7 537 151
BC013145.1 - 4 - 5 450 151
D86965.1 - 5 - 6 1937 151

View Full Product Details