Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TM4SF20
Entrez Gene ID
79853
Gene Name
transmembrane 4 L six family member 20
Gene Aliases
PRO994, SLI5, TCCE518
Chromosome Location
Chr.2: 227362041 - 227379306 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_024795.3 NP_079071.2 3 - 4 440 81
XM_011511876.2 XP_011510178.1 3 - 4 346 81
GenBank mRNA AK026453.1 - 3 - 4 439 81
AY358671.1 - 3 - 4 432 81
BC035754.1 - 3 - 4 427 81
BC137256.1 - 3 - 4 426 81
BC137257.1 - 3 - 4 426 81
DA926038.1 - 3 - 4 440 81

View Full Product Details