Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- OR7E47P
- Entrez Gene ID
-
26628
- Gene Name
- olfactory receptor family 7 subfamily E member 47 pseudogene
- Gene Aliases
-
OR7E141
- Chromosome Location
-
Chr.12: 52079696 - 52108319 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_120439.1 |
|
1 - 2 |
86 |
78 |
View Full Product Details