The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000902.3 | NP_000893.2 | 16 - 17 | 1722 | 105 |
NM_007287.2 | NP_009218.2 | 16 - 17 | 1698 | 105 | |
NM_007288.2 | NP_009219.2 | 16 - 17 | 1744 | 105 | |
NM_007289.2 | NP_009220.2 | 16 - 17 | 1789 | 105 | |
XM_006713646.3 | XP_006713709.1 | 16 - 17 | 1736 | 105 | |
XM_006713647.3 | XP_006713710.1 | 16 - 17 | 4882 | 105 | |
XM_011512855.2 | XP_011511157.1 | 16 - 17 | 1813 | 105 | |
XM_011512856.2 | XP_011511158.1 | 16 - 17 | 1740 | 105 | |
XM_011512857.2 | XP_011511159.1 | 16 - 17 | 1711 | 105 | |
GenBank mRNA | AK291761.1 | - | 16 - 17 | 1813 | 105 |
BC101632.1 | - | 16 - 17 | 1680 | 105 | |
BC101658.1 | - | 16 - 17 | 1678 | 105 | |
BC143465.1 | - | 16 - 17 | 1680 | 105 | |
BX647134.1 | - | 16 - 17 | 1693 | 105 | |
J03779.1 | - | 15 - 16 | 1613 | 105 | |
X07166.1 | - | 15 - 16 | 1598 | 105 | |
Y00811.1 | - | 16 - 17 | 1737 | 105 |