The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000902.3 | NP_000893.2 | 6 - 7 | 655 | 140 |
NM_007287.2 | NP_009218.2 | 6 - 7 | 631 | 140 | |
NM_007288.2 | NP_009219.2 | 6 - 7 | 677 | 140 | |
NM_007289.2 | NP_009220.2 | 6 - 7 | 722 | 140 | |
XM_006713646.3 | XP_006713709.1 | 6 - 7 | 669 | 140 | |
XM_006713647.3 | XP_006713710.1 | 6 - 7 | 3815 | 140 | |
XM_011512855.2 | XP_011511157.1 | 6 - 7 | 746 | 140 | |
XM_011512856.2 | XP_011511158.1 | 6 - 7 | 673 | 140 | |
XM_011512857.2 | XP_011511159.1 | 6 - 7 | 644 | 140 | |
GenBank mRNA | AK291761.1 | - | 6 - 7 | 746 | 140 |
BC101632.1 | - | 6 - 7 | 613 | 140 | |
BC101658.1 | - | 6 - 7 | 611 | 140 | |
BC143465.1 | - | 6 - 7 | 613 | 140 | |
BX647134.1 | - | 6 - 7 | 627 | 139 | |
J03779.1 | - | 5 - 6 | 546 | 140 | |
X07166.1 | - | 5 - 6 | 531 | 140 | |
Y00811.1 | - | 6 - 7 | 670 | 140 |