The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001145099.1 | NP_001138571.1 | 5 - 6 | 870 | 57 |
NM_017585.3 | NP_060055.2 | 5 - 6 | 870 | 57 | |
XM_011518189.2 | XP_011516491.2 | 5 - 6 | 847 | 57 | |
XM_017014236.1 | XP_016869725.1 | 5 - 6 | 852 | 57 | |
GenBank mRNA | AB209108.1 | - | 3 - 4 | 1760 | 57 |
AJ011372.2 | - | 5 - 6 | 812 | 57 | |
AK074836.1 | - | 5 - 6 | 820 | 57 | |
AK074927.1 | - | 5 - 6 | 853 | 57 | |
AK222919.1 | - | 5 - 6 | 815 | 57 | |
AK225106.1 | - | 5 - 6 | 743 | 57 | |
BC013740.1 | - | 5 - 6 | 804 | 57 | |
Y17803.1 | - | 5 - 6 | 821 | 57 |