The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015869.4 | NP_056953.2 | 1 - 2 | 176 | 92 |
XM_011533842.2 | XP_011532144.1 | 1 - 2 | 2209 | 92 | |
XM_011533843.2 | XP_011532145.1 | 1 - 2 | 2204 | 92 | |
GenBank mRNA | AB451337.1 | - | 1 - 2 | 85 | 92 |
AB451486.1 | - | 1 - 2 | 85 | 92 | |
AJ563369.1 | - | 1 - 2 | 45 | 92 | |
BI820841.1 | - | 1 - 2 | 211 | 92 | |
D83233.1 | - | 1 - 2 | 243 | 92 | |
U63415.1 | - | 1 - 2 | 85 | 92 | |
U79012.1 | - | 1 - 2 | 175 | 92 |