The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001064.3 | NP_001055.1 | 12 - 13 | 1743 | 99 |
NM_001135055.2 | NP_001128527.1 | 12 - 13 | 1743 | 99 | |
NM_001258028.1 | NP_001244957.1 | 13 - 14 | 1767 | 99 | |
NR_047580.1 | 12 - 13 | 1633 | 99 | ||
XM_011534054.1 | XP_011532356.1 | 13 - 14 | 1705 | 99 | |
XM_011534055.1 | XP_011532357.1 | 13 - 14 | 1792 | 99 | |
GenBank mRNA | AK057092.1 | - | 6 - 7 | 1572 | 99 |
AK092084.1 | - | 7 - 8 | 2606 | 99 | |
AK093667.1 | - | 11 - 12 | 1402 | 99 | |
AK223614.1 | - | 12 - 13 | 1651 | 99 | |
AK289454.1 | - | 12 - 13 | 1619 | 99 | |
AK293438.1 | - | 13 - 14 | 1675 | 99 | |
AK301231.1 | - | 12 - 13 | 1541 | 99 | |
AK303191.1 | - | 13 - 14 | 1610 | 99 | |
BC002433.2 | - | 13 - 14 | 1532 | 99 | |
BC008615.1 | - | 7 - 8 | 887 | 99 | |
BC009970.2 | - | 12 - 13 | 1641 | 99 | |
BC024026.2 | - | 10 - 11 | 1312 | 99 | |
BQ430230.1 | - | 3 - 4 | 291 | 99 | |
BX647248.1 | - | 12 - 13 | 1629 | 99 | |
CA421169.1 | - | 2 - 3 | 99 | ||
EU794667.1 | - | 12 - 13 | 1641 | 99 | |
GQ472197.1 | - | 7 - 8 | 887 | 99 | |
KU178361.1 | - | 12 - 13 | 1571 | 99 | |
KU178363.1 | - | 5 - 6 | 457 | 99 | |
L12711.1 | - | 12 - 13 | 1669 | 99 | |
U55017.1 | - | 12 - 13 | 1571 | 99 | |
X67688.1 | - | 13 - 14 | 1641 | 99 |