The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001135055.2 | NP_001128527.1 | 14 - 15 | 2052 | 87 |
XM_011534054.1 | XP_011532356.1 | 15 - 16 | 2014 | 87 | |
GenBank mRNA | BX647248.1 | - | 14 - 15 | 1938 | 87 |