The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_156039.3 | NP_724781.1 | 16 - 17 | 2621 | 57 |
XM_011540748.2 | XP_011539050.1 | 15 - 16 | 2146 | 57 | |
XM_011540749.1 | XP_011539051.1 | 16 - 17 | 2618 | 57 | |
XM_011540750.1 | XP_011539052.1 | 11 - 12 | 1451 | 57 | |
XM_017000370.1 | XP_016855859.1 | 15 - 16 | 2183 | 57 | |
GenBank mRNA | M59820.1 | - | 16 - 17 | 2206 | 57 |