The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_032864.3 | NP_116253.2 | 7 - 8 | 950 | 68 |
GenBank mRNA | AK027842.1 | - | 6 - 7 | 524 | 68 |
AK092038.1 | - | 7 - 8 | 933 | 68 | |
BC000777.2 | - | 5 - 6 | 535 | 68 | |
BC063655.1 | - | 5 - 6 | 592 | 68 | |
BC105004.1 | - | 7 - 8 | 791 | 68 | |
BC105006.1 | - | 7 - 8 | 791 | 68 |