Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
KIF27
Entrez Gene ID
55582
Gene Name
kinesin family member 27
Gene Aliases
-
Chromosome Location
Chr.9: 83832795 - 83921539 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001271927.1 NP_001258856.1 3 - 4 678 94
NM_001271928.1 NP_001258857.1 3 - 4 678 94
NM_017576.2 NP_060046.1 3 - 4 678 94
XM_011518848.2 XP_011517150.1 3 - 4 679 94
XM_011518849.2 XP_011517151.1 4 - 5 795 94
XM_011518850.2 XP_011517152.1 3 - 4 768 94
XM_011518854.2 XP_011517156.1 4 - 5 898 94
XM_011518856.2 XP_011517158.1 4 - 5 898 94
XM_017014900.1 XP_016870389.1 5 - 6 947 94
XM_017014901.1 XP_016870390.1 5 - 6 829 94
XM_017014902.1 XP_016870391.1 4 - 5 875 94
XM_017014903.1 XP_016870392.1 4 - 5 898 94
XM_017014904.1 XP_016870393.1 4 - 5 795 94
XM_017014905.1 XP_016870394.1 4 - 5 898 94
XM_017014906.1 XP_016870395.1 4 - 5 898 94
XM_017014907.1 XP_016870396.1 4 - 5 898 94
XM_017014908.1 XP_016870397.1 4 - 5 898 94
XM_017014909.1 XP_016870398.1 3 - 4 500 94
XM_017014910.1 XP_016870399.1 4 - 5 795 94
XM_017014914.1 XP_016870403.1 3 - 4 752 94
GenBank mRNA AK297689.1 - 3 - 4 635 94
AY237536.1 - 3 - 4 640 94
AY237537.1 - 3 - 4 640 94
AY237538.1 - 3 - 4 640 94
BC140788.1 - 3 - 4 639 94

View Full Product Details