The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001173452.1 | NP_001166923.1 | 3 - 4 | 1066 | 140 |
NM_001173453.1 | NP_001166924.1 | 3 - 4 | 1066 | 140 | |
NM_005653.4 | NP_005644.2 | 3 - 4 | 1066 | 140 | |
GenBank mRNA | AK291264.1 | - | 3 - 4 | 717 | 140 |
AK308087.1 | - | 3 - 4 | 417 | 140 | |
BC003634.2 | - | 3 - 4 | 804 | 140 | |
M84810.1 | - | 3 - 4 | 708 | 140 | |
U03494.1 | - | 3 - 4 | 1036 | 140 | |
U03495.1 | - | 4 - 5 | 853 | 140 |