Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TFCP2
Entrez Gene ID
7024
Gene Name
transcription factor CP2
Gene Aliases
LBP1C, LSF, LSF1D, SEF, TFCP2C
Chromosome Location
Chr.12: 51093756 - 51173143 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001173452.1 NP_001166923.1 5 - 6 1285 83
NM_005653.4 NP_005644.2 5 - 6 1285 83
GenBank mRNA AK291264.1 - 5 - 6 936 83
AK308087.1 - 5 - 6 636 83
BC003634.2 - 5 - 6 1023 83

View Full Product Details