The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_138962.3 | NP_620412.1 | 11 - 12 | 964 | 135 |
GenBank mRNA | AK297295.1 | - | 7 - 8 | 648 | 135 |
BC001526.2 | - | 11 - 12 | 964 | 135 |