The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001242837.1 | NP_001229766.1 | 7 - 8 | 1032 | 92 |
XM_011519929.1 | XP_011518231.1 | 7 - 8 | 789 | 92 | |
XM_011519930.2 | XP_011518232.1 | 7 - 8 | 993 | 92 | |
GenBank mRNA | AK027891.1 | - | 7 - 8 | 941 | 92 |
AK223558.1 | - | 7 - 8 | 960 | 92 |