The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001289000.1 | NP_001275929.1 | 10 - 11 | 1260 | 115 |
NM_001289001.1 | NP_001275930.1 | 10 - 11 | 1270 | 115 | |
NM_153609.3 | NP_705837.1 | 10 - 11 | 1339 | 115 | |
XM_011529989.2 | XP_011528291.1 | 7 - 8 | 966 | 115 | |
GenBank mRNA | AJ319876.1 | - | 9 - 10 | 1202 | 115 |
AY055384.1 | - | 10 - 11 | 1339 | 115 | |
AY358398.1 | - | 10 - 11 | 1270 | 115 | |
CR456446.1 | - | 10 - 11 | 1260 | 115 |