The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001270585.1 | NP_001257514.1 | 1 - 2 | 63 | 64 |
XM_011511864.1 | XP_011510166.1 | 1 - 2 | 61 | 64 | |
XM_011511866.1 | XP_011510168.1 | 1 - 2 | 61 | 64 | |
XM_017004960.1 | XP_016860449.1 | 1 - 2 | 61 | 64 | |
GenBank mRNA | AK054711.1 | - | 1 - 2 | 62 | 64 |
AK295566.1 | - | 1 - 2 | 60 | 64 | |
HY127810.1 | - | 1 - 2 | 64 | 64 |