The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002293.3 | NP_002284.3 | 11 - 12 | 2249 | 72 |
GenBank mRNA | J03202.1 | - | 11 - 12 | 2251 | 72 |