The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002293.3 | NP_002284.3 | 13 - 14 | 2653 | 89 |
GenBank mRNA | CF125114.1 | - | 1 - 2 | 148 | 89 |
J03202.1 | - | 13 - 14 | 2655 | 89 |