The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002293.3 | NP_002284.3 | 20 - 21 | 3825 | 64 |
GenBank mRNA | BC015586.2 | - | 2 - 3 | 182 | 64 |
J03202.1 | - | 20 - 21 | 3827 | 64 |