The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002293.3 | NP_002284.3 | 22 - 23 | 4104 | 77 |
GenBank mRNA | BC015586.2 | - | 4 - 5 | 461 | 77 |
J03202.1 | - | 22 - 23 | 4106 | 77 |