The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002293.3 | NP_002284.3 | 7 - 8 | 1688 | 77 |
GenBank mRNA | CN388644.1 | - | 5 - 6 | 600 | 77 |
J03202.1 | - | 7 - 8 | 1690 | 77 |