Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
LAMC1
Entrez Gene ID
3915
Gene Name
laminin subunit gamma 1
Gene Aliases
LAMB2
Chromosome Location
Chr.1: 183023460 - 183145592 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_002293.3 NP_002284.3 8 - 9 120
GenBank mRNA BI768612.1 - 3 - 4 356 120
J03202.1 - 8 - 9 120

View Full Product Details