The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001297669.1 | NP_001284598.1 | 8 - 9 | 1543 | 90 |
NM_001297670.1 | NP_001284599.1 | 7 - 8 | 1433 | 90 | |
NM_001297671.1 | NP_001284600.1 | 7 - 8 | 1169 | 90 | |
NM_001297672.1 | NP_001284601.1 | 7 - 8 | 1169 | 90 | |
NM_015149.4 | NP_055964.3 | 8 - 9 | 1544 | 90 | |
XM_011509339.2 | XP_011507641.1 | 9 - 10 | 1590 | 90 | |
XM_011509341.1 | XP_011507643.1 | 8 - 9 | 1497 | 90 | |
XM_011509342.2 | XP_011507644.1 | 8 - 9 | 1497 | 90 | |
XM_017000756.1 | XP_016856245.1 | 7 - 8 | 1018 | 90 | |
GenBank mRNA | AB023176.1 | - | 7 - 8 | 1109 | 90 |
AF186779.1 | - | 7 - 8 | 1130 | 90 | |
AF186780.1 | - | 8 - 9 | 1506 | 90 | |
AK295179.1 | - | 7 - 8 | 1307 | 90 | |
AK295611.1 | - | 4 - 5 | 746 | 90 | |
AK304293.1 | - | 7 - 8 | 1094 | 90 | |
AK311701.1 | - | 7 - 8 | 1015 | 90 | |
AK316526.1 | - | 8 - 9 | 1481 | 90 | |
AL080117.1 | - | 6 - 7 | 912 | 90 | |
BC136591.1 | - | 8 - 9 | 1248 | 90 | |
BX537416.1 | - | 8 - 9 | 1484 | 90 |