The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001105251.2 | NP_001098721.1 | 1 - 2 | 136 | 84 |
XM_017010091.1 | XP_016865580.1 | 1 - 2 | 140 | 84 | |
GenBank mRNA | AB002303.1 | - | 1 - 2 | 76 | 84 |