Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CENPF
Entrez Gene ID
1063
Gene Name
centromere protein F
Gene Aliases
CENF, CILD31, PRO1779, STROMS, hcp-1
Chromosome Location
Chr.1: 214603181 - 214664574 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_016343.3 NP_057427.3 10 - 11 1617 113
XM_011509082.2 XP_011507384.1 10 - 11 1625 113
XM_017000086.1 XP_016855575.1 10 - 11 1604 113
GenBank mRNA AK307690.1 - 10 - 11 1600 113
U19769.1 - - 1613 113
U30872.1 - 10 - 11 1515 113

View Full Product Details