Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CENPF
Entrez Gene ID
1063
Gene Name
centromere protein F
Gene Aliases
CENF, CILD31, PRO1779, STROMS, hcp-1
Chromosome Location
Chr.1: 214603181 - 214664574 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_016343.3 NP_057427.3 1 - 2 127 101
XM_011509082.2 XP_011507384.1 1 - 2 135 101
GenBank mRNA AK307690.1 - 1 - 2 110 101
BU662228.1 - 1 - 2 154 101
U19769.1 - - 123 101

View Full Product Details