The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001190996.1 | NP_001177925.1 | 6 - 7 | 869 | 62 |
NM_006409.3 | NP_006400.2 | 6 - 7 | 871 | 62 | |
GenBank mRNA | AB209059.1 | - | 6 - 7 | 804 | 62 |
AK223015.1 | - | 5 - 6 | 695 | 62 | |
BC039594.1 | - | 6 - 7 | 834 | 62 | |
BC047889.1 | - | 6 - 7 | 804 | 62 | |
BC054027.1 | - | 6 - 7 | 803 | 62 | |
EU668326.1 | - | 6 - 7 | 857 | 62 | |
GQ472234.1 | - | 5 - 6 | 695 | 62 | |
Y08999.1 | - | 5 - 6 | 743 | 62 |